2,4,6-Trimethylphenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

B24060.03 B24060.06 B24060.14
B24060.03EA 38 GBP
B24060.03 B24060.06 B24060.14
2,4,6-Trimethylphenylboronic acid ≥97%
2,4,6-Trimethylphenylboronic acid

Purity: 97% CAS No.: 5980-97-2 Molecular Formula: C9H13BO2 Formula Weight: 164.01 Melting Point: ca 120-127° Beilstein Reference: 2831927

Formula: C₉H₁₃BO₂
MW: 164,01 g/mol
Melting Pt: ∼120…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00236060
CAS Number: 5980-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR