3-Methylbenzyl alcohol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(Hydroxymethyl)toluole, (3-Methylphenyl)methanol, m-Tolylmethanol
B20875.14
B20875.22
B20875.14EA
58
GBP
B20875.14
B20875.22
3-Methylbenzyl alcohol ≥98%
3-Methylbenzyl alcohol
Purity: 98%
CAS No.: 587-03-1
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 105°(221°F)
Refractive Index: 1.5340
Boiling Point: 216-217°
Beilstein Reference: 2324516
MDL No.: MFCD00004646
Formula:
C₈H₁₀O MW: 122,17 g/mol Boiling Pt: 216…217 °C Melting Pt: < –20 °C Density: 1,013 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004646 CAS Number: 587-03-1 EINECS: 209-595-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...