2,5-Xylenol, Supelco®

Supplier: Merck
Danger

Synonyms: 2,5-Dimethylphenol, 2-Hydroxy-p-xylene, 2-Hydroxy-4-xylene

M-Clarity™ quality level = MQ100

36714-1G
36714-1GEA 33.6 GBP
36714-1G
2,5-Xylenol, Supelco®
2,5-Xylenol
Formula: C₈H₁₀O
MW: 122,17 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1,169 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002237
CAS Number: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=734bb91d1e32c2ae88997124a3f4a702d2987f33&vid=734bb91d1e32c2ae88997124a3f4a702d2987f33&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR