3-(Trifluoromethyl)benzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3-(Trifluoromethyl)phenyl)methanol, 3-(Hydroxymethyl)benzotrifluoride

A16739.06 A16739.14
A16739.06EA 38.3 GBP
A16739.06 A16739.14
3-(Trifluoromethyl)benzyl alcohol ≥97%
3-(Trifluoromethyl)benzyl alcohol

Purity: 97% CAS No.: 349-75-7 Molecular Formula: C8H7F3O Formula Weight: 176.14 Flash Point: 84°(183°F) Refractive Index: 1.4600 Boiling Point: 79-80°/3mm Beilstein Reference: 1450161 MDL No.: MFCD00004645

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 79…80 °C (3 mmHg)
Density: 1.293
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004645
CAS Number: 349-75-7
EINECS: 206-489-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR