4-Chlorobenzophenone ≥99%
Supplier: Thermo Scientific
Synonyms:
p-Chlorobenzophenone
A17458.0E
A17458.22
A17458.36
A17458.0EEA
284
GBP
A17458.0E
A17458.22
A17458.36
4-Chlorobenzophenone ≥99%
4-Chlorobenzophenone
Purity: 99%
CAS No.: 134-85-0
Molecular Formula: C13H9ClO
Formula Weight: 216.67
Flash Point: 143°(289°F)
Melting Point: 75-77°
Boiling Point: 332°
Beilstein Reference: 512043
MDL No.: MFCD00000622
Formula:
C₁₃H₉ClO MW: 216,67 g/mol Boiling Pt: 332 °C Melting Pt: 75…77 °C Flash Pt: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000622 CAS Number: 134-85-0 EINECS: 205-160-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...