Bromocresol purple

Supplier: RAL (REACTIFS RAL SA)
Warning

Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide5',5''-Dibromo-o-cresolsulphonphthalein

720-1086EA 97.1 GBP
720-1086
Bromocresol purple
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
MW: 540,23 g/mol
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR