o-Toluidine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-Toluidine, 2-Aminotoluene, 2-Methylaniline

M-Clarity™ quality level = MQ200

8.08312.0500
8.08312.0500EA 106 GBP
8.08312.0500
o-Toluidine for synthesis, Sigma-Aldrich®
o-Toluidine
Formula: C₇H₉N
MW: 107,16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: < –15 °C
Density: 0,998 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS Number: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1,II
Merck Index: 12,09674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR