4-Hexylbenzoic acid ≥99%

Supplier: Thermo Scientific

A14168.03 A14168.06
A14168.03EA 46.1 GBP
A14168.03 A14168.06
4-Hexylbenzoic acid ≥99%
4-Hexylbenzoic acid

Purity: 99% CAS No.: 21643-38-9 Molecular Formula: C13H18O2 Formula Weight: 206.29 Melting Point: (C-N) 97-99° Beilstein Reference: 2046454

Formula: C₁₃H₁₈O₂
MW: 206,28 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00013997
CAS Number: 21643-38-9
EINECS: 244-490-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR