3'-Hydroxyacetophenone 98
Supplier: Thermo Fisher Scientific
|
Warning
|
A12547.22
A12547.36
A12547.22EA
40.8
GBP
A12547.22
A12547.36
3'-Hydroxyacetophenone 98
3'-Hydroxyacetophenone
Purity: 98%
CAS No.: 121-71-1
Molecular Formula: C8H8O2
Formula Weight: 136.15
Melting Point: 93-97°
Boiling Point: 296°
MDL No.: MFCD00002298
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 296 °C Melting Pt: 93…97 °C Density: 1.1 Storage Temperature: Ambient |
MDL Number:
MFCD00002298 CAS Number: 121-71-1 EINECS: 204-494-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...