2-Acetylbenzoic acid 99%

Supplier: Thermo Fisher Scientific
Warning

102470050 102470250 102471000
ACRO102470050EA 49.1 GBP
ACRO102470050 ACRO102470250 ACRO102471000
2-Acetylbenzoic acid 99%
2-Acetylbenzoic acid
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002475
CAS Number: 577-56-0
EINECS: 209-413-2

Order Now

Specification Test Results

Appearance Light yellow to light orange Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR