α-Bromo-2-iodotoulene ≥96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: α-Bromo-o-iodotoulene, 2-Iodobenzyl bromide, 1-(Bromomethyl)-2-iodobenzene

B21715.14 B21715.22
B21715.14EA 0 GBP
B21715.14 B21715.22
α-Bromo-2-iodotoulene ≥96%
α-Bromo-2-iodotoulene

Purity: 96% CAS No.: 40400-13-3 Molecular Formula: C7H6BrI Formula Weight: 296.94 Melting Point: 53-58° Beilstein Reference: 2079736 UN No.: 3261 MDL No.: MFCD00236046

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 279
Melting Pt: 55.5
MDL Number: MFCD00236046
CAS Number: 40400-13-3
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR