1-Bromo-4-propylbenzene ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: (4-Bromophenyl)propane, 1-Bromo-4-n-propylbenzene

H26236.06 H26236.14 H26236.22
H26236.06EA 26.5 GBP
H26236.06 H26236.14 H26236.22
1-Bromo-4-propylbenzene ≥99%
1-Bromo-4-propylbenzene

Purity: 99% CAS No.: 588-93-2 Molecular Formula: C9H11Br Formula Weight: 199.09 Flash Point: 95°(203°F) Refractive Index: 1.537 Boiling Point: 225° MDL No.: MFCD00012456

Formula: C₉H₁₁Br
MW: 199,09 g/mol
Boiling Pt: 225 °C
Melting Pt: –41,5 °C
Density: 1,286
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012456
CAS Number: 588-93-2
EINECS: 454-790-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR