Anisole, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: MethoxybenzeneMethyl phenyl ether

M-Clarity™ quality level = MQ200

8.19243.6190EA 0 GBP
8.19243.6190
Anisole, Sigma-Aldrich®
Anisole
Formula: C₇H₈O
MW: 108,14 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: –37 °C
Density: 0,9961 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
Merck Index: 13,00672
https://ni.vwr.com/stibo/web/std.lang.all/96/93/759693.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR