O-Phenylenediamine 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD
A11946.30
A11946.0B
A11946.0I
A11946.14
A11946.30EA
39.8
GBP
A11946.30
A11946.0B
A11946.0I
A11946.14
O-Phenylenediamine 98%
o-Phenylenediamine
Purity: 98%
CAS No.: 95-54-5
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 136°(277°F)
Physical Form: Crystals/flakes
Density: 1.270
Sensitivity: Air and Light Sensitive
Melting Point: 100-103°
Boiling Point: 256-258°
Solubility: Soluble in alcohol, chloroform, ether, slightly soluble in water
Beilstein Reference: 606074
Merck Reference: 14,7284
UN No.: 1673
MDL No.: MFCD00007721
Formula:
C₆H₈N₂ MW: 108,14 g/mol Boiling Pt: 256…258 °C Melting Pt: 100…103 °C Density: 1,270 Flash Pt: 136 °C (277 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007721 CAS Number: 95-54-5 EINECS: 202-430-6 UN: 1673 ADR: 6.1,III Merck Index: 14,7284 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...