Sulphanilamide, Supelco®

Supplier: Merck

Synonyms: 4-Aminobenzenesulphonamide

M-Clarity™ quality level = MQ200

46874-250MG
46874-250MGEA 42.4 GBP
46874-250MG
Sulphanilamide, Supelco®
Sulphanilamide
Formula: H₂NC₆H₄SO₂NH₂
MW: 172,21 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 163…167 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5cfa0cc6050793333a7b5f6b1e5ab0986202cdf&vid=f5cfa0cc6050793333a7b5f6b1e5ab0986202cdf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR