Pentafluorothiophenol ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
B22323.06
B22323.14
B22323.22
B22323.06EA
75.6
GBP
B22323.06
B22323.14
B22323.22
Pentafluorothiophenol ≥97%
Pentafluorothiophenol
Purity: 97%
CAS No.: 771-62-0
Molecular Formula: C6HF5S
Formula Weight: 200.13
Flash Point: 51°(123°F)
Refractive Index: 1.4650
Odour: Stench
Melting Point: -24°
Boiling Point: 143°
Beilstein Reference: 1876292
UN No.: 2920
MDL No.: MFCD00004828
Formula:
C₆HF₅S Boiling Pt: 143 °C Melting Pt: –24 °C Density: 1,572 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004828 CAS Number: 771-62-0 EINECS: 212-236-3 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...