2-(Trifluoromethoxy)thiophenol ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 2-(Trifluoromethoxy)benzene-1-thiol, 2-(Trifluoromethoxy)benzenethiol

L19911.03 L19911.06
L19911.03EA 69.7 GBP
L19911.03 L19911.06
2-(Trifluoromethoxy)thiophenol ≥99%
2-(Trifluoromethoxy)thiophenol

Purity: 99% CAS No.: 175278-01-0 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4768 Boiling Point: 53-55°/15mm UN No.: 2810 MDL No.: MFCD00052320

Formula: C₇H₅F₃OS
MW: 194,18 g/mol
Boiling Pt: 53…55 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052320
CAS Number: 175278-01-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR