4,5-Difluoro-2-nitroaniline

Warning

Synonyms: 3,4-Difluoro-6-nitroaniline

CFF1044.1 CFF1044.5 CFF1044.25
CHMPCFF1044.1EA 0 GBP
CHMPCFF1044.1 CHMPCFF1044.5 CHMPCFF1044.25
4,5-Difluoro-2-nitroaniline
4,5-Difluoro-2-nitroaniline
Formula: C₆H₄F₂N₂O₂
MW: 174,11 g/mol
MDL Number: MFCD00010876
CAS Number: 78056-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR