2,3-Difluoro-4-hydroxybenzoic acid ≥96%

Supplier: Thermo Fisher Scientific
Warning

H26087.MD H26087.03
H26087.MDEA 0 GBP
H26087.MD H26087.03
2,3-Difluoro-4-hydroxybenzoic acid ≥96%
2,3-Difluoro-4-hydroxybenzoic acid

Purity: 99% CAS No.: 175968-39-5 Molecular Formula: C7H4F2O3 Formula Weight: 174.10 Melting Point: 205-210° MDL No.: MFCD01321116

Formula: C₇H₄F₂O₃
Melting Pt: 205…210 °C
Storage Temperature: Ambient
MDL Number: MFCD01321116
CAS Number: 175968-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR