3-Amino-p-anisic acid ≥98%

Supplier: Thermo Scientific
Warning

B20669.06 B20669.14
B20669.06EA 29.6 GBP
B20669.06 B20669.14
3-Amino-p-anisic acid ≥98%
3-Amino-p-anisic acid

Purity: 98+% CAS No.: 2840-26-8 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 208-210° MDL No.: MFCD00002521

Formula: C₈H₉NO₃
MW: 167,16 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002521
CAS Number: 2840-26-8
EINECS: 220-634-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR