2-Cyanobenzoic acid 96%

Supplier: Thermo Scientific
Warning

H50237.03 H50237.06
H50237.03EA 45.1 GBP
H50237.03 H50237.06
2-Cyanobenzoic acid 96%
2-Cyanobenzoic acid

Purity: 96% CAS No.: 3839-22-3 Molecular Formula: C8H5NO2 Formula Weight: 147.13 Melting Point: 210-212° MDL No.: MFCD00045796

Formula: C₈H₅NO₂
MW: 147,13 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00045796
CAS Number: 3839-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR