Dibutyl phthalate-[ring-D₄] (98% D)
Supplier: LGC Standards PROMOCHEM
|
Danger
|
Synonyms:
Dibutyl phthalate-[ring-²H₄]
CILDLM13670.1
CILDLM13670.25
PROOCILDLM13670.1EA
1020
GBP
PROOCILDLM13670.1
PROOCILDLM13670.25
Dibutyl phthalate-[ring-D₄] (98% D)
Dibutyl phthalate-[ring-D4]
Formula:
C₁₆H₁₈D₄O₄ Storage Temperature: Ambient |
CAS Number:
93952-11-5 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...