3-Aminobenzamide 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: m-Aminobenzamide, INO-1001

339080010 339080050 339080250
ACRO339080010EA 58 GBP
ACRO339080010 ACRO339080050 ACRO339080250
3-Aminobenzamide 98%
3-Aminobenzamide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 341
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00007989
CAS Number: 3544-24-9
EINECS: 222-586-9

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR