Benzene-1,3,5-tricarbonyl trichloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1,3,5-Benzenetricarboxylic acid chloride, 1,3,5-Benzenetricarbonyl chloride, Trimesoylchloride, Trimesoyl chloride
L07255.06
L07255.14
L07255.22
L07255.06EA
26.9
GBP
L07255.06
L07255.14
L07255.22
Benzene-1,3,5-tricarbonyl trichloride ≥98%
Benzene-1,3,5-tricarbonyl trichloride
Purity: 98+%
CAS No.: 4422-95-1
Molecular Formula: C9H3Cl3O3
Formula Weight: 265.48
Flash Point:
Formula:
C₉H₃Cl₃O₃ MW: 265,48 g/mol Boiling Pt: 180 °C (16 mmHg) Melting Pt: 34…38 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000679 CAS Number: 4422-95-1 EINECS: 224-594-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...