2,4,5-Trichlorophenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L11551.03
L11551.06
L11551.03EA
0
GBP
L11551.03
L11551.06
2,4,5-Trichlorophenyl isothiocyanate ≥98%
2,4,5-Trichlorophenyl isothiocyanate
Purity: 98%
CAS No.: 23165-46-0
Molecular Formula: C7H2Cl3NS
Formula Weight: 238.52
Odour: Irritating
Melting Point: 42-47°
Boiling Point: 141-142°/1mm
Beilstein Reference: 881221
UN No.: 2923
MDL No.: MFCD00041052
Formula:
C₇H₂Cl₃NS MW: 238.52 g/mol Boiling Pt: 141…142 °C (1 mmHg) Melting Pt: 42…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041052 CAS Number: 23165-46-0 EINECS: 245-471-5 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...