1-(2-Bromoethoxy)-4-nitrobenzene 98%
Supplier: Thermo Scientific
|
Warning
|
385290050
385290250
ACRO385290050EA
71.3
GBP
ACRO385290050
ACRO385290250
1-(2-Bromoethoxy)-4-nitrobenzene 98%
1-(2-Bromoethoxy)-4-nitrobenzene
Formula:
C₈H₈BrNO₃ MW: 246,06 g/mol Melting Pt: 62…65 °C |
MDL Number:
MFCD00031363 CAS Number: 13288-06-7 |
Specification Test Results
Appearance | Yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 62 to 65 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...