2-Bromo-5-iodobenzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

L19247.03 L19247.06
L19247.03EA 0 GBP
L19247.03 L19247.06
2-Bromo-5-iodobenzoic acid ≥97%
2-Bromo-5-iodobenzoic acid

Purity: 97% CAS No.: 25252-00-0 Molecular Formula: C7H4BrIO2 Formula Weight: 326.92 Melting Point: 174-176° MDL No.: MFCD00079716

Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00079716
CAS Number: 25252-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR