2-Phenylbenzimidazole ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Phenyl-1H-benzimidazole, 2-Phenyl-1H-benzo[d]imidazole

A10996.06 A10996.14 A10996.22
A10996.06EA 22.9 GBP
A10996.06 A10996.14 A10996.22
2-Phenylbenzimidazole ≥97%
2-Phenylbenzimidazole

Purity: 97% CAS No.: 716-79-0 Molecular Formula: C13H10N2 Formula Weight: 194.24 Melting Point: 296-297° Beilstein Reference: 7087 Merck Reference: 14,7274 MDL No.: MFCD00005592

Formula: C₁₃H₁₀N₂
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: 296…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00005592
CAS Number: 716-79-0
EINECS: 211-939-2
Merck Index: 14,7274

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR