4-Hydroxy-1-methylpiperidine
Supplier: Molekula
|
Warning
|
Synonyms:
1-Methylpiperidin-4-ol
, 1-Methyl-4-piperidinol
, N-Methyl-4-piperidinol
MOLEM79427424EA
0
GBP
MOLEM79427424
4-Hydroxy-1-methylpiperidine
4-Hydroxy-1-methylpiperidine
Formula:
C₆H₁₃NO MW: 115,18 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: 29…31 °C Density: 0,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006500 CAS Number: 106-52-5 EINECS: 203-406-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...