2-Piperidone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: δ-Valerolactame, delta-Valerolactam, Piperidin-2-one

A12271.14 A12271.22
A12271.14EA 65.8 GBP
A12271.14 A12271.22
2-Piperidone ≥98%
2-Piperidone

Purity: 98+% CAS No.: 675-20-7 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 147°(296°F) Melting Point: 35-39° Boiling Point: 255-257° Beilstein Reference: 106434 MDL No.: MFCD00006037

Formula: C₅H₉NO
MW: 99,13 g/mol
Boiling Pt: 255…257 °C
Melting Pt: 35…39 °C
Density: 1,073 g/cm³ (40 °C)
Flash Pt: 147 °C (296 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006037
CAS Number: 675-20-7
EINECS: 211-622-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR