6-Phenylcaproic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Phenylhexanoic acid

A11187.03 A11187.06
A11187.03EA 0 GBP
A11187.03 A11187.06
6-Phenylcaproic acid ≥98%
6-Phenylcaproic acid

Purity: 98+% CAS No.: 5581-75-9 Molecular Formula: C6H5(CH2)5CO2H Formula Weight: 192.26 Flash Point: 110°(230°F) Refractive Index: 1.5110 Melting Point: 17-19° Boiling Point: 157-160°/0.3mm Beilstein Reference: 1950106 MDL No.: MFCD00014381

Formula: C₁₂H₁₆O₂
MW: 192,26 g/mol
Boiling Pt: 157…160 °C (0,3 mmHg)
Melting Pt: 17…19 °C
Density: 1,030
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014381
CAS Number: 5581-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR