Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
|
Warning
|
Synonyms:
3-Methyl-1-butanol
, Isopentyl alcohol
, 3-Methylbutanol
, iso-Pentanol
20797.292EA
50.5
GBP
20797.292
20797.326
Isoamyl alcohol, TECHNICAL
Isoamyl alcohol
Formula:
(CH₃)₂CHCH₂CH₂OH MW: 88,15 g/mol Boiling Pt: 130 °C (1013 hPa) Melting Pt: –117 °C Density: 0,812 g/cm³ (20 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...