3-Methylacetylacetone mixture of tautomers 95%

Supplier: Thermo Fisher Scientific
Warning

L01135.06 L01135.14
L01135.06EA 31 GBP
L01135.06 L01135.14
3-Methylacetylacetone mixture of tautomers 95%
3-Methylacetylacetone

Purity: 95% CAS No.: 815-57-6 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 56°(132°F) Refractive Index: 1.4420 Boiling Point: 170-171° Beilstein Reference: 635909 UN No.: 1224 MDL No.: MFCD00008762

Formula: C₆H₁₀O₂
Boiling Pt: 170…171 °C
Melting Pt: –31 °C
Density: 0,980
Flash Pt: 56 °C (132 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008762
CAS Number: 815-57-6
EINECS: 212-420-3
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR