Pivalaldehyde (trimethylacetaldehyde) 96%

Supplier: Thermo Scientific
Danger

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

368360050 368360250 368361000
ACRO368360050EA 118 GBP
ACRO368360050 ACRO368360250 ACRO368361000
Pivalaldehyde (trimethylacetaldehyde) 96%
Pivalaldehyde (Trimethylacetaldehyde)
Formula: C₅H₁₀O
MW: 86,13 g/mol
Boiling Pt: 74 °C (1013 hPa)
Melting Pt: 6 °C
Density: 0,775 g/cm³
Flash Pt: –15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006962
CAS Number: 630-19-3
EINECS: 211-134-6
UN: 1989

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3800 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR