1,12-Dodecanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,12-Dihydroxy dodecane
M-Clarity™ quality level = MQ200
8.03501.0100
8.03501.1000
8.03501.0100EA
103
GBP
8.03501.0100
8.03501.1000
1,12-Dodecanediol for synthesis, Sigma-Aldrich®
1,12-Dodecanediol
Formula:
C₁₂H₂₆O₂ MW: 202,34 g/mol Boiling Pt: 313 °C (1013 hPa) Melting Pt: 78…79 °C Density: 1,08 g/cm³ (20 °C) |
MDL Number:
MFCD00004755 CAS Number: 5675-51-4 EINECS: 227-133-9 |
Specification Test Results
Solubility |
|
Melting Point | 79 °C |
Molar Mass | 202.33 g/mol |
Boiling Point | 324 °C (1013 hPa) |
Flash Point | 176 °C |
Ignition Point | 300 °C |
Synonyms | 1,12-Dihydroxydodecane, Dodecamethylene glycol |
Hill Formula | C12H26O2 |
Chemical Formula | HO(CH2)12OH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...