1,12-Diaminododecane ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N12N, Dodecamethylendiamine, 1,12-Dodecanediamine
L04258.06
L04258.14
L04258.06EA
30.5
GBP
L04258.06
L04258.14
1,12-Diaminododecane ≥98%
1,12-Diaminododecane
Purity: 98+%
CAS No.: 2783-17-7
Molecular Formula: C12H28N2
Formula Weight: 200.37
Flash Point: 155°(311°F)
Melting Point: 68-72°
Beilstein Reference: 1742765
UN No.: 3259
MDL No.: MFCD00008155
Formula:
C₁₂H₂₈N₂ MW: 200,37 g/mol Boiling Pt: 275 °C (1013 hPa) Melting Pt: 68…72 °C Flash Pt: 155 °C (311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008155 CAS Number: 2783-17-7 EINECS: 220-489-6 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...