2-(2-Aminoethoxy)ethanol 98%
Supplier: Thermo Scientific
|
Danger
|
L18897.22
L18897.36
L18897.22EA
35.9
GBP
L18897.22
L18897.36
2-(2-Aminoethoxy)ethanol 98%
2-(2-Aminoethoxy)ethanol
Purity: 98%
CAS No.: 929-06-6
Molecular Formula: H2NCH2CH2OCH2CH2OH
Formula Weight: 105.14
Flash Point: 127°(260°F)
Refractive Index: 1.4570
Melting Point: -11°
Boiling Point: 125-131°/23mm
Beilstein Reference: 906728
UN No.: 3055
MDL No.: MFCD00008181
Formula:
C₄H₁₁NO₂ MW: 105,14 g/mol Boiling Pt: 125…131 °C (23 mmHg) Melting Pt: –11 °C Density: 1,057 Flash Pt: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008181 CAS Number: 929-06-6 EINECS: 213-195-4 UN: 3055 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...