1-Decyl isothiocyanate ≥96%
Supplier: Thermo Scientific
Synonyms:
Decyl isothiocyanate
L12165.06
L12165.03
L12165.06EA
0
GBP
L12165.06
L12165.03
1-Decyl isothiocyanate ≥96%
1-Decyl isothiocyanate
Purity: 96%
CAS No.: 24540-94-1
Molecular Formula: CH3(CH2)9NCS
Formula Weight: 199.36
Refractive Index: 1.4860
Odour: Irritating
Boiling Point: 170-171°/35mm
Beilstein Reference: 6288335
UN No.: 1760
MDL No.: MFCD00041120
Formula:
C₁₁H₂₁NS MW: 199,36 g/mol Boiling Pt: 170…171 °C (35 mmHg) Density: 0,89 g/cm³ (20 °C) |
MDL Number:
MFCD00041120 CAS Number: 24540-94-1 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...