Iodoacetamide 98%

Supplier: Thermo Fisher Scientific
Danger

122270050 122270250 122271000 122275000
ACRO122270050EA 54.3 GBP
ACRO122270050 ACRO122270250 ACRO122271000 ACRO122275000
Iodoacetamide 98%
Iodoacetamide
Formula: ICH₂CONH₂
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: >90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point ≥90 °C
Titration Argentometric ≥97.5 % (ex I)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR