Ethyl difluoroacetate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Difluoroacetic ethyl ester
A11215.06
A11215.14
A11215.22
A11215.06EA
56.9
GBP
A11215.06
A11215.14
A11215.22
Ethyl difluoroacetate ≥98%
Ethyl difluoroacetate
Purity: 98%
CAS No.: 454-31-9
Molecular Formula: F2CHCO2CH2CH3
Formula Weight: 124.09
Flash Point: 25°(77°F)
Refractive Index: 1.3475
Boiling Point: 98-99°
Beilstein Reference: 1748597
UN No.: 2924
MDL No.: MFCD00013578
Formula:
C₄H₆F₂O₂ MW: 124,09 g/mol Boiling Pt: 98…99 °C Density: 1,242 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013578 CAS Number: 454-31-9 EINECS: 207-223-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...