D-Methionine
Supplier: Molekula
Synonyms:
D(+)-Methionine
, (R)-(+)-2-Amino-4-(methylthio)butyric acid
, H-D-Met-OH
, (R)-2-Amino-4-(methylthio)butanoic acid
, (R)-(+)-2-Amino-4-(methylthio)butanoic acid
, (R)-(+)-Methionine
MOLE30263029-25GEA
0
GBP
MOLE30263029-25G
D-Methionine
D-Methionine
Formula:
C₅H₁₁NO₂S MW: 149,21 g/mol Boiling Pt: 278 °C (1013 hPa) Melting Pt: 136 °C |
MDL Number:
MFCD00002622 CAS Number: 348-67-4 EINECS: 206-483-6 Merck Index: 13,06004 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...