Sodium butyrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Butyric acid sodium salt
, Sodium Butyrate
M-Clarity™ quality level = MQ200
8.17500.9025EA
0
GBP
8.17500.9025
8.17500.9999
Sodium butyrate, Sigma-Aldrich®
Sodium butyrate
Formula:
C₄H₇NaO₂ MW: 110,09 g/mol Melting Pt: 244 °C Density: 1,324 g/cm³ (30 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002816 CAS Number: 156-54-7 EINECS: 205-857-6 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 244 °C |
Molar Mass | 110.09 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...