Ethyl myristate ≥98%
Supplier: Thermo Scientific
Synonyms:
Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester
A11646.AE
A11646.AP
A11646.AEEA
39.5
GBP
A11646.AE
A11646.AP
Ethyl myristate ≥98%
Ethyl myristate
Purity: 98%
CAS No.: 124-06-1
Molecular Formula: C16H32O2
Formula Weight: 256.43
Flash Point: 135°(275°F)
Refractive Index: 1.4360
Odour: Weak
Melting Point: 12°
Boiling Point: 295°
Beilstein Reference: 1776382
Merck Reference: 14,6333
MDL No.: MFCD00008984
Formula:
C₁₆H₃₂O₂ MW: 256,43 g/mol Boiling Pt: 295 °C Melting Pt: 12 °C Density: 0,860 Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008984 CAS Number: 124-06-1 EINECS: 204-675-4 Merck Index: 14,6333 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...