Methyl valerate, Supelco®
Supplier: Merck
Synonyms:
Valeric acid methyl ester, Methyl Pentanoate
M-Clarity™ quality level = MQ100
94560-1ML
94560-5ML
94560-1MLEA
46.7
GBP
94560-1ML
94560-5ML
Methyl valerate, Supelco®
Methyl valerate
Formula:
C₆H₁₂O₂ MW: 116,16 g/mol Boiling Pt: 126,5 °C (1013 hPa) Melting Pt: –91 °C Density: 0,89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009478 CAS Number: 624-24-8 EINECS: 210-838-0 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...