Ethyl valerate 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl Pentanoate, Valeric acid ethyl ester
410470050
410471000
410475000
ACRO410470050EA
43.4
GBP
ACRO410470050
ACRO410471000
ACRO410475000
Ethyl valerate 99%
Ethyl valerate
Formula:
C₇H₁₄O₂ MW: 130,19 g/mol Boiling Pt: 145 °C Melting Pt: –91 °C Density: 0,87 g/cm³ Flash Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009479 CAS Number: 539-82-2 EINECS: 208-726-1 UN: 3272 ADR: 3,III Merck Index: 12,10042 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3990 to 1.4010 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...