N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)

Supplier: PanReac AppliChem

Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid

A1060.1000
A1060.1000EA 1900 GBP
A1060.1000
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182,2 g/mol
Melting Pt: 270…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=058001d050ee2844850d66215657dd9a3095cfce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR