Hexanoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

B23027.22 B23027.36
B23027.22EA 39.2 GBP
B23027.22 B23027.36
Hexanoyl chloride ≥98%
Hexanoyl chloride

Purity: 98% CAS No.: 142-61-0 Molecular Formula: CH3(CH2)4COCl Formula Weight: 134.61 Flash Point: 50°(122°F) Refractive Index: 1.4260 Odour: Acrid Melting Point: -87° Boiling Point: 150-153° Merck Reference: 14,1763 UN No.: 2920 MDL No.: MFCD00000760

Formula: C₆H₁₁ClO
Boiling Pt: 150…153 °C
Melting Pt: –87 °C
Density: 0,971
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000760
CAS Number: 142-61-0
EINECS: 205-549-1
UN: 2920
ADR: 8,II
Merck Index: 14,1763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR