Propiononitrile 99%, extra pure
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Propionitrile
180895000
180890025
180891000
ACRO180895000EA
85.1
GBP
ACRO180895000
ACRO180890025
ACRO180891000
Propiononitrile 99%, extra pure
Propiononitrile
Formula:
C₃H₅N MW: 55,08 g/mol Boiling Pt: 97 °C (760 mmHg) Melting Pt: –93 °C Density: 0,77 g/cm³ Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001948 CAS Number: 107-12-0 EINECS: 203-464-4 UN: 2404 ADR: 3,II Merck Index: 12,08012 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3650 to 1.3670 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...