5-Bromovaleric acid 97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Bromopentanoic acid

169820100 169820500 169822500
ACRO169820100EA 33.8 GBP
ACRO169820100 ACRO169820500 ACRO169822500
5-Bromovaleric acid 97%
5-Bromovaleric acid
Formula: BrH₂C(CH₂)₃COOH
MW: 181,03 g/mol
Boiling Pt: 142…145 °C (13 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004414
CAS Number: 2067-33-6
EINECS: 218-185-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 37 to 41 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR