Methyl-5-bromovalerate 97%
Supplier: Thermo Scientific
Synonyms:
Methyl 5-Bromovalerate, 5-Bromovaleric acid methyl ester
126460100
126460250
ACRO126460100EA
119
GBP
ACRO126460100
ACRO126460250
Methyl-5-bromovalerate 97%
Methyl 5-bromovalerate
Formula:
BrH₂C(CH₂)₃COOCH₃ MW: 195,06 g/mol Boiling Pt: 79…80 °C (5,33 hPa) Density: 1,36 g/cm³ Flash Pt: 99 °C |
MDL Number:
MFCD00000265 CAS Number: 5454-83-1 EINECS: 226-709-7 |
Specification Test Results
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4620 to 1.4640 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...